Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 2-cyanoacetate 95%

Supplier: AMBEED, INC
Warning

A831123-100G A831123-1G A831123-25G A831123-5G
76993-500EA 215 USD InStock
76993-500 76993-502 76993-504 76993-506
Sodium 2-cyanoacetate 95%
Sodium 2-cyanoacetate
Precaución: for Research Use Only


Formula: C₃H₂NNaO₂
MDL Number: MFCD00067028
CAS Number: 1071-36-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty