Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-2H-indazole-6-boronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A834669-1G A834669-250MG A834669-5G
77036-600EA 66 USD InStock
77036-600 77036-602 77036-604
2-Methyl-2H-indazole-6-boronic acid pinacol ester 98%
2-Methyl-2H-indazole-6-boronic acid pinacol ester


Formula: C₁₄H₁₉BN₂O₂
MDL Number: MFCD11109435
CAS Number: 1204580-79-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty