Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Methoxyphenyl)-2-butanone 97%

Supplier: AMBEED, INC
Warning

A837601-100G A837601-25G A837601-500G
77036-610EA 47 USD InStock
77036-610 77036-612 77036-614
4-(4-Methoxyphenyl)-2-butanone 97%
4-(4-Methoxyphenyl)-2-butanone
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 72
Melting Pt: 10
MDL Number: MFCD00008791
CAS Number: 104-20-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty