Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Phenyl-2-(di-t-butylphosphino)indol 98%

Supplier: AMBEED, INC
Warning

A838494-1G A838494-250MG
76993-518EA 110 USD InStock
76993-518 76993-520
N-Phenyl-2-(di-t-butylphosphino)indol 98%
N-Phenyl-2-(di-t-butylphosphino)indol
Precaución: for Research Use Only


Formula: C₂₂H₂₈NP
MDL Number: MFCD06798302
CAS Number: 740815-37-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty