Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Bis(tert-butyl)-2-methoxybenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A843210-1G A843210-250MG A843210-25G A843210-5G
77036-648EA 55 USD InStock
77036-648 77036-650 77036-652 77036-654
3,5-Bis(tert-butyl)-2-methoxybenzaldehyde 97%
3,5-Bis(tert-butyl)-2-methoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₁₆H₂₄O₂
MDL Number: MFCD02093684
CAS Number: 135546-15-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty