Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methyl-2-indoleboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A843909-1G A843909-250MG A843909-5G
77019-644EA 218 USD InStock
77019-644 77019-646 77019-648
5-Methyl-2-indoleboronic acid pinacol ester 98%
5-Methyl-2-indoleboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₅H₂₀BNO₂
MW: 257.1358 g/mol
MDL Number: MFCD11858361
CAS Number: 1256358-93-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty