Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-3-methylbenzyl alcohol 98+%

Supplier: AMBEED, INC
Warning

A846858-1G A846858-250MG A846858-5G
77019-660EA 20 USD InStock
77019-660 77019-662 77019-664
2-Methoxy-3-methylbenzyl alcohol 98+%
2-Methoxy-3-methylbenzyl alcohol
Precaución: for Research Use Only


Formula: C₉H₁₂O₂
MW: 152.19 g/mol
MDL Number: MFCD12031989
CAS Number: 74090-48-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty