Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-3-(3,4-methylenedioxyphenyl)propionaldehyde 98%

Supplier: AMBEED, INC
Warning

A850017-100G A850017-25G A850017-500G A850017-5G
77006-810EA 48 USD InStock
77006-810 77006-812 77006-814 77006-816
2-Methyl-3-(3,4-methylenedioxyphenyl)propionaldehyde 98%
2-Methyl-3-(3,4-methylenedioxyphenyl)propionaldehyde
Precaución: for Research Use Only


Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
MDL Number: MFCD00067053
CAS Number: 1205-17-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty