Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Methioninamide hydrochloride 98%

Supplier: AMBEED, INC
Warning

A850839-1G A850839-25G A850839-5G
77006-818EA 10 USD InStock
77006-818 77006-820 77006-822
L-Methioninamide hydrochloride 98%
L-Methioninamide hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₃ClN₂OS
MDL Number: MFCD00039084
CAS Number: 16120-92-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty