Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethylaminoacetic acid hydrochloride 97%

Supplier: AMBEED, INC
Warning

A854037-10G A854037-250MG
77036-690EA 326 USD InStock
77036-690 77036-692
Diethylaminoacetic acid hydrochloride 97%
Diethylaminoacetic acid hydrochloride


Formula: C₆H₁₄ClNO₂
MDL Number: MFCD00050569
CAS Number: 24376-01-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty