Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-formylbenzonitrile 95%

Supplier: AMBEED, INC
Warning

A854977-100MG A854977-1G A854977-250MG A854977-5G
76993-610EA 23 USD InStock
76993-610 76993-612 76993-614 76993-616
2-Bromo-3-formylbenzonitrile 95%
2-Bromo-3-formylbenzonitrile
Precaución: for Research Use Only


Formula: C₈H₄BrNO
MDL Number: MFCD18206376
CAS Number: 446864-55-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty