Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethoxy-4-methylbenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A857773-100MG A857773-250MG A857773-50MG A857773-1G
76993-630EA 128 USD InStock
76993-630 76993-632 76993-634 77308-460
3,5-Dimethoxy-4-methylbenzaldehyde 95%
3,5-Dimethoxy-4-methylbenzaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₃
MDL Number: MFCD12025247
CAS Number: 1011-27-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty