Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethoxy)isophthalonitrile 95+%

Supplier: AMBEED, INC
Warning

A861734-1G A861734-250MG A861734-5G
76993-658EA 7 USD InStock
76993-658 76993-660 76993-662
4-(Trifluoromethoxy)isophthalonitrile 95+%
4-(Trifluoromethoxy)isophthalonitrile
Precaución: for Research Use Only


Formula: C₉H₃F₃N₂O
MDL Number: MFCD08459281
CAS Number: 1020063-01-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty