Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Methylenanthic acid 98%

Supplier: AMBEED, INC
Warning

A861963-100G A861963-25G A861963-5G
77006-892EA 282 USD InStock
77006-892 77006-894 77006-896
(±)-2-Methylenanthic acid 98%
(±)-2-Methylenanthic acid
Precaución: for Research Use Only


Formula: C₈H₁₆O₂
MDL Number: MFCD00142944
CAS Number: 1188-02-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty