Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Dimethylamino)-4-methylphenol 95%

Supplier: AMBEED, INC
Warning

A863659-100MG A863659-1G A863659-250MG
77036-726EA 43 USD InStock
77036-726 77036-728 77036-730
3-(Dimethylamino)-4-methylphenol 95%
3-(Dimethylamino)-4-methylphenol
Precaución: for Research Use Only


Formula: C₉H₁₃NO
MDL Number: MFCD08361738
CAS Number: 119-31-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty