Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-1-(4-(trifluoromethyl)phenyl)ethanone hydrochloride 97%

Supplier: AMBEED, INC
Warning

A865364-100MG A865364-1G A865364-250MG
76993-674EA 15 USD InStock
76993-674 76993-676 76993-678
2-Amino-1-(4-(trifluoromethyl)phenyl)ethanone hydrochloride 97%
2-Amino-1-(4-(trifluoromethyl)phenyl)ethanone hydrochloride
Precaución: for Research Use Only


Formula: C₉H₉ClF₃NO
MW: 239.62 g/mol
MDL Number: MFCD11974299
CAS Number: 339-58-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty