Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Ethoxyanilino)-4-oxobutanoic acid 97%

Supplier: AMBEED, INC
Warning

A865887-1G A865887-250MG A865887-5G
77006-912EA 65 USD InStock
77006-912 77006-914 77006-916
4-(4-Ethoxyanilino)-4-oxobutanoic acid 97%
4-(4-Ethoxyanilino)-4-oxobutanoic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₅NO₄
MDL Number: MFCD00020528
CAS Number: 59256-45-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty