Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-Ethyl-1H-pyrazol-4-yl)methylamine 95%

Supplier: AMBEED, INC
Danger

A879349-100MG A879349-1G A879349-250MG A879349-5G
77036-776EA 17 USD InStock
77036-776 77036-778 77036-780 77674-310
(1-Ethyl-1H-pyrazol-4-yl)methylamine 95%
(1-Ethyl-1H-pyrazol-4-yl)methylamine
Precaución: for Research Use Only


Formula: C₆H₁₁N₃
MDL Number: MFCD02253772
CAS Number: 856696-09-8
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty