Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-N,N-dimethylethane-1-sulfonamide hydrochloride 95%

Supplier: AMBEED, INC
Warning

A881987-1G A881987-250MG
77019-828EA 241 USD InStock
77019-828 77019-830
2-Amino-N,N-dimethylethane-1-sulfonamide hydrochloride 95%
2-Amino-N,N-dimethylethane-1-sulfonamide hydrochloride
Precaución: for Research Use Only


Formula: C₄H₁₂N₂O₂S·ClH
MDL Number: MFCD04112919
CAS Number: 91893-69-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty