Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-chloro-4-hydroxy-1-methyl-2-oxo-1,2-dihydroquinoline-3-carboxylate 97%

Supplier: AMBEED, INC
Warning

A882329-100MG A882329-1G A882329-250MG
77019-832EA 10 USD InStock
77019-832 77019-834 77019-836
Methyl 5-chloro-4-hydroxy-1-methyl-2-oxo-1,2-dihydroquinoline-3-carboxylate 97%
Methyl 5-chloro-4-hydroxy-1-methyl-2-oxo-1,2-dihydroquinoline-3-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₀ClNO₄
MW: 267.66 g/mol
MDL Number: MFCD09032505
CAS Number: 637027-41-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty