Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Hydroxy-2-methyl-3,4-dihydroisoquinolin-1(2H)-one 95%

Supplier: AMBEED, INC
Warning

A886828-100MG A886828-250MG
77019-854EA 48 USD InStock
77019-854 77019-856
7-Hydroxy-2-methyl-3,4-dihydroisoquinolin-1(2H)-one 95%
7-Hydroxy-2-methyl-3,4-dihydroisoquinolin-1(2H)-one
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO₂
MDL Number: MFCD24719603
CAS Number: 913613-99-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty