Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[1,2,4]Triazol-4-ylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A891756-100MG A891756-1G A891756-250MG A891756-5G
77006-982EA 17 USD InStock
77006-982 77006-984 77006-986 77006-988
4-[1,2,4]Triazol-4-ylbenzoic acid 98%
4-[1,2,4]Triazol-4-ylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₇N₃O₂
MDL Number: MFCD06654839
CAS Number: 157069-48-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty