Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(4-(Dimethylamino)benzylidene)-2-thioxothiazolidin-4-one 92% + (mixture of isomers)

Supplier: AMBEED, INC
Warning

A895713-100G A895713-1G A895713-25G A895713-5G
77036-820EA 52 USD InStock
77036-820 77036-822 77036-824 77036-826
5-(4-(Dimethylamino)benzylidene)-2-thioxothiazolidin-4-one 92% + (mixture of isomers)
5-(4-(Dimethylamino)benzylidene)-2-thioxothiazolidin-4-one
Precaución: for Research Use Only


Formula: C₁₂H₁₂N₂OS₂
MDL Number: MFCD04969049
CAS Number: 536-17-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty