Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,3S)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid 95%

Supplier: AMBEED, INC
Warning

A897887-100MG A897887-250MG
76993-824EA 76 USD InStock
76993-824 76993-826
(1S,3S)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid 95%
(1S,3S)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₉NO₄
MDL Number: MFCD02259726
CAS Number: 161601-29-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty