Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

NS-1619 98%

Supplier: AMBEED, INC
Warning

A900983-10MG A900983-1MG A900983-5MG
77036-840EA 22 USD InStock
77036-840 77036-842 77036-844
NS-1619 98%
NS-1619
Precaución: for Research Use Only


Formula: C₁₅H₈F₆N₂O₂
MDL Number: MFCD00928190
CAS Number: 153587-01-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty