Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(tert-Butylaminocarbonylmethyl)phenylboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A902496-100MG A902496-1G A902496-250MG A902496-5G
77019-910EA 37 USD InStock
77019-910 77019-912 77019-914 77019-916
4-(tert-Butylaminocarbonylmethyl)phenylboronic acid pinacol ester 97%
4-(tert-Butylaminocarbonylmethyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₈H₂₈BNO₃
MDL Number: MFCD16618971
CAS Number: 1256359-83-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty