Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Di-tert-butyl-5-nitrophenyl methyl carbonate 97%

Supplier: AMBEED, INC
Danger

A913990-1G A913990-250MG A913990-5G
76993-870EA 44 USD InStock
76993-870 76993-872 76993-874
2,4-Di-tert-butyl-5-nitrophenyl methyl carbonate 97%
2,4-Di-tert-butyl-5-nitrophenyl methyl carbonate
Precaución: for Research Use Only


Formula: C₁₆H₂₃NO₅
MW: 309.35 g/mol
MDL Number: MFCD11113067
CAS Number: 873055-55-1
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty