Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-tert-Butyl (6-oxopiperidin-3-yl)carbamate 95%

Supplier: AMBEED, INC
Warning

A914279-100MG A914279-1G A914279-250MG
77036-876EA 17 USD InStock
77036-876 77036-878 77036-880
(R)-tert-Butyl (6-oxopiperidin-3-yl)carbamate 95%
(R)-tert-Butyl (6-oxopiperidin-3-yl)carbamate
Precaución: for Research Use Only


Formula: C₁₀H₁₈N₂O₃
MW: 214.26 g/mol
MDL Number: MFCD13195282
CAS Number: 1228566-94-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty