Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-(chloromethyl)pyridine hydrochloride 95%

Supplier: AMBEED, INC
Danger

A915795-250MG A915795-5G
76993-876EA 90 USD InStock
76993-876 76993-878
3-Chloro-2-(chloromethyl)pyridine hydrochloride 95%
3-Chloro-2-(chloromethyl)pyridine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₆Cl₃N
MW: 198.48 g/mol
MDL Number: MFCD20527153
CAS Number: 124425-87-2
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty