Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl thiazol-5-ylcarbamate 97%

Supplier: AMBEED, INC
Warning

A918708-100MG A918708-10G A918708-1G A918708-250MG A918708-5G
77036-896EA 11 USD InStock
77036-896 77036-898 77036-900 77036-902 77036-904
tert-Butyl thiazol-5-ylcarbamate 97%
tert-Butyl thiazol-5-ylcarbamate
Precaución: for Research Use Only


Formula: C₈H₁₂N₂O₂S
MDL Number: MFCD11977627
CAS Number: 942631-50-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty