Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-5-methoxyquinazoline 97%

Supplier: AMBEED, INC
Warning

A921504-100MG A921504-1G A921504-250MG
77036-910EA 13 USD InStock
77036-910 77036-912 77036-914
2,4-Dichloro-5-methoxyquinazoline 97%
2,4-Dichloro-5-methoxyquinazoline
Precaución: for Research Use Only


Formula: C₉H₆Cl₂N₂O
MW: 229.06 g/mol
MDL Number: MFCD10697887
CAS Number: 61948-59-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty