Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-methoxy-5-nitropyrimidine 97%

Supplier: AMBEED, INC
Warning

A922062-1G A922062-250MG A922062-5G A922062-100MG
77036-918EA 343 USD InStock
77036-918 77036-920 77036-922 77670-356
2-Chloro-4-methoxy-5-nitropyrimidine 97%
2-Chloro-4-methoxy-5-nitropyrimidine
Precaución: for Research Use Only


Formula: C₅H₄ClN₃O₃
MW: 189.55 g/mol
MDL Number: MFCD00955634
CAS Number: 282102-07-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty