Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Aminooxy)propan-2-ol hydrochloride 95%

Supplier: AMBEED, INC
Danger

A941071-100MG A941071-1G A941071-250MG A941071-5G
77020-034EA 45 USD InStock
77020-034 77020-036 77020-038 77020-040
1-(Aminooxy)propan-2-ol hydrochloride 95%
1-(Aminooxy)propan-2-ol hydrochloride
Precaución: for Research Use Only


Formula: C₃H₁₀ClNO₂
MDL Number: MFCD27920818
CAS Number: 950595-72-9
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty