Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methylindole-6-carboxylic acid methyl ester 97%

Supplier: AMBEED, INC
Warning

A950555-1G A950555-250MG A950555-25G A950555-5G
77007-178EA 8 USD InStock
77007-178 77007-180 77007-182 77007-184
N-Methylindole-6-carboxylic acid methyl ester 97%
N-Methylindole-6-carboxylic acid methyl ester
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₂
MW: 189.22 g/mol
MDL Number: MFCD06245515
CAS Number: 1204-32-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty