Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Aminomethyl)-1-methyl-1H-indazole 95%

Supplier: AMBEED, INC
Danger

A951015-100MG A951015-1G A951015-250MG
77020-064EA 35 USD InStock
77020-064 77020-066 77020-068
5-(Aminomethyl)-1-methyl-1H-indazole 95%
5-(Aminomethyl)-1-methyl-1H-indazole
Precaución: for Research Use Only


Formula: C₉H₁₁N₃
MDL Number: MFCD12028599
CAS Number: 267413-27-4
UN: 3259
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty