Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butyl({[2-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl})amine 96%

Supplier: AMBEED, INC
Warning

A967368-1G A967368-250MG A967368-5G
77020-134EA 87 USD InStock
77020-134 77020-136 77020-138
Butyl({[2-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl})amine 96%
Butyl({[2-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl})amine
Precaución: for Research Use Only


Formula: C₁₇H₂₈BNO₂
MDL Number: MFCD14280318
CAS Number: 1256359-08-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty