Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-Fluoro-[1,1'-biphenyl]-4-amine 97+%

Supplier: AMBEED, INC
Warning

A970225-100MG A970225-1G A970225-250MG A970225-5G
77037-038EA 19 USD InStock
77037-038 77037-040 77037-042 77037-044
3'-Fluoro-[1,1'-biphenyl]-4-amine 97+%
3'-Fluoro-[1,1'-biphenyl]-4-amine
Precaución: for Research Use Only


Formula: C₁₂H₁₀FN
MW: 187.21 g/mol
MDL Number: MFCD03424688
CAS Number: 5728-66-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty