Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cinnarizine 99+%

Supplier: AMBEED, INC
Danger

A971851-100MG A971851-25MG
77007-208EA 6 USD InStock
77007-208 77007-210
Cinnarizine 99+%
Cinnarizine
Precaución: for Research Use Only


Formula: C₂₆H₂₈N₂
MDL Number: MFCD00056037
CAS Number: 298-57-7
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty