Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2',3',5'-Tri-O-acetyl-2-amino-6-chloropurine riboside 98+%

Supplier: AMBEED, INC
Warning

A986690-1G A986690-250MG A986690-25G A986690-5G
77007-258EA 44 USD InStock
77007-258 77007-260 77007-262 77007-264
2',3',5'-Tri-O-acetyl-2-amino-6-chloropurine riboside 98+%
2',3',5'-Tri-O-acetyl-2-amino-6-chloropurine riboside
Precaución: for Research Use Only


Formula: C₁₆H₁₈ClN₅O₇
MDL Number: MFCD03425473
CAS Number: 16321-99-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty