Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(-)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane ≥96%, ee 99%

Supplier: AMBEED, INC
Warning

A992160-100G A992160-10G A992160-1G A992160-250MG A992160-25G A992160-5G
77007-278EA 235 USD InStock
77007-278 77007-280 77007-282 77007-284 77007-286 77007-288
(-)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane ≥96%, ee 99%
(-)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane
Precaución: for Research Use Only


Formula: C₃₁H₃₀O₄
MW: 466.58 g/mol
MDL Number: MFCD00064467
CAS Number: 93379-48-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty