Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-o-anisaldehyde 98%

Supplier: AMBEED, INC
Warning

A180674-25G A180674-100G A180674-10G A180674-500G
77083-414EA 14 USD InStock
77083-414 77274-058 77274-060 77274-062
5-Bromo-o-anisaldehyde 98%
5-Bromo-o-anisaldehyde
Precaución: for Research Use Only


Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 269
Melting Pt: 117
MDL Number: MFCD00003347
CAS Number: 25016-01-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty