Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dibromo-1-fluorobenzene 98%

Supplier: AMBEED, INC
Warning

A207930-25G A207930-1000G A207930-100G A207930-10G A207930-500G
77086-822EA 10 USD InStock
77086-822 77287-858 77287-860 77287-862 77287-864 77287-866
3,5-Dibromo-1-fluorobenzene 98%
3,5-Dibromo-1-fluorobenzene
Precaución: for Research Use Only


Formula: C₆H₃Br₂F
MW: 253.9 g/mol
Boiling Pt: 204
MDL Number: MFCD00061119
CAS Number: 1435-51-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty