Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Benzodioxole 98%

Supplier: AMBEED, INC
Warning

A210984-25G A210984-1000G A210984-100G A210984-500G A210984-5G
77087-146EA 18 USD InStock
77087-146 77192-632 77192-634 77192-636 77192-638
1,3-Benzodioxole 98%
1,3-Benzodioxole
Precaución: for Research Use Only


Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 172
Melting Pt: -18
MDL Number: MFCD00005818
CAS Number: 274-09-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty