Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dibromopyridine 98%

Supplier: AMBEED, INC
Warning

A222663-25G A222663-1000G A222663-100G A222663-10G A222663-500G
77088-482EA 11 USD InStock
77088-482 77275-508 77275-510 77275-512 77275-514
3,5-Dibromopyridine 98%
3,5-Dibromopyridine
Precaución: for Research Use Only


Formula: C₅H₃Br₂N
MDL Number: MFCD00014634
CAS Number: 625-92-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty