Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-mercapto-1,3,4-thiadiazole 97%

Supplier: AMBEED, INC
Warning

A237933-25G A237933-100G A237933-500G A237933-1000G
77090-068EA 8 USD InStock
77090-068 77113-164 77113-166 77113-168 77113-170 77685-662
2-Amino-5-mercapto-1,3,4-thiadiazole 97%
2-Amino-5-mercapto-1,3,4-thiadiazole
Precaución: for Research Use Only


Formula: C₂H₃N₃S₂
MDL Number: MFCD00003108
CAS Number: 2349-67-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty