Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-(dimethylamino)acrylate 97%

Supplier: AMBEED, INC
Warning

A239076-25G A239076-1000G A239076-100G A239076-10G A239076-500G
77090-194EA 6 USD InStock
77090-194 77113-200 77113-202 77113-204 77113-206
Ethyl-3-(dimethylamino)acrylate 97%
Ethyl-3-(dimethylamino)acrylate
Precaución: for Research Use Only


Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
MDL Number: MFCD00144269
CAS Number: 924-99-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty