Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-Pro-OH 98%

Supplier: AMBEED, INC
Warning

A246166-25G A246166-1000G A246166-100G A246166-10G A246166-500G
77090-800EA 6 USD InStock
77090-800 77276-178 77276-180 77276-182 77276-184
Boc-Pro-OH 98%
Boc-Pro-OH
Precaución: for Research Use Only


Formula: C₁₀H₁₇NO₄
MDL Number: MFCD00037324
CAS Number: 15761-39-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty