Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Octyl-1-dodecanol 97%

Supplier: AMBEED, INC
Warning

A264125-25G A264125-100G A264125-10G A264125-500G
77092-206EA 8 USD InStock
77092-206 77276-620 77276-622 77276-624
2-Octyl-1-dodecanol 97%
2-Octyl-1-dodecanol
Precaución: for Research Use Only


Formula: C₂₀H₄₂O
MDL Number: MFCD01310428
CAS Number: 5333-42-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty