Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nazartinib ≥98%

Supplier: AMBEED, INC
Warning

A100729-1MG A100729-2MG A100729-5MG
77093-482EA 32 USD InStock
77093-482 77093-484 77093-486
Nazartinib ≥98%
Nazartinib
Precaución: for Research Use Only


Formula: C₂₆H₃₁lN₆O₂
MDL Number: MFCD28963918
CAS Number: 1508250-71-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty