Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Diethyl-4-iodobenzamide 98%

Supplier: AMBEED, INC
Warning

A104768-100MG A104768-1G A104768-250MG A104768-5G
77093-660EA 29 USD InStock
77093-660 77093-662 77093-664 77093-666
N,N-Diethyl-4-iodobenzamide 98%
N,N-Diethyl-4-iodobenzamide
Precaución: for Research Use Only


Formula: C₁₁H₁₄INO
MW: 303.13 g/mol
MDL Number: MFCD02217872
CAS Number: 77350-52-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty